Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P2C01MV01

Millipore

Pellicon® 2 Mini Ultrafiltration Module

filtration area 0.1 m2, NMWCO 1000 kDa

Synonym(s):

Pellicon® 2 Mini Ultrafiltration Module PLCXK V 0.1 m2

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,966.00

$1,966.00


Estimated to ship onMarch 31, 2025Details



Select a Size

Change View
1 EA
$1,966.00

About This Item

UNSPSC Code:
23151806

$1,966.00


Estimated to ship onMarch 31, 2025Details


material

composite regenerated cellulose (CRC)
polypropylene screen
polyurethane adhesive

Quality Level

manufacturer/tradename

Pellicon®

parameter

3.5 bar max. transmembrane pressure (50 psig) at 30 °C
5-35 L/min recirculation rate at 0.4 bar (6 psi)

technique(s)

analytical sample preparation: suitable
protein purification: suitable

H

2.16 cm (0.85 in.)

L

21 cm (8.3 in.)

W

5.6 cm (2.2 in.)

filtration area

0.1 m2

membrane nominal area

0.1 m2

gravimetric extractables

≤1500 mg/m2

matrix

Ultracel®

pore size

1000 kDa NMWCO

pH

2-13

Looking for similar products? Visit Product Comparison Guide

General description

The Pellicon® 2 Ultrafiltration PLCXK Mini Cassette sets the standard in cassette-style tangential flow filtration devices. P2C01MV01 was designed with process scale-up and applications development in mind.

Millipore′s PLCXK polyethersulfone membrane has void-free structures that guard against leakage of solutes through microdefects normally associated with voids beneath the thin skins of conventional UF membranes. These void-free membranes are more permeable, resulting in high-flux with equivalent or superior product retention. These void-free membranes provide the advantages of fast, high yield processing and smaller systems.

The Pellicon® 2 Mini Ultrafiltration Module feature a filtration area of 0.1 m2 with a pore size of 1000 kDa NMWCO.

Application

Concentration, desalting or buffer exchange of:
  • Protein solutions
  • Polysaccharide solutions
  • Virus suspensions


Harvest, washing or clarification of:
  • Cell cultures and lysates
  • Colloidal suspensions
  • Viral cultures

Features and Benefits

  • Type V screen (open channel)
  • Leading-edge void- free membranes to match virtually any separation challenge
  • Short flow path for higher flux and higher resolution separation capability
  • Choice of flow channel configuration providing process optimization capability
  • Predictable, fast, scale-up
  • True linear scalability from laboratory size modules to industrial assemblies for processing thousands of liters
  • Brings higher resolution, improved yields and superior back-pressure resistance
  • Ultracel membranes are composite membranes cast on a microporous substrate for defect-free membranes with superior

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.
Prefiltration Rquirements: 100 µm

Analysis Note

Gravimetric Extractables: after a water flush

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service