Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OX0165

Millipore

Oleic Acid

OmniPur® Grade

Synonym(s):

OmniPur® Oleic Acid, Octadecenoic Acid, 9-Octadecenoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
282.47
MDL number:
UNSPSC Code:
12352211
EC Index Number:
204-007-1

vapor pressure

<0.1 hPa ( 20 °C)

form

liquid

bp

360 °C/1013 hPa

mp

16 °C

transition temp

flash point 180 °C

density

~0.89 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9+

InChI key

ZQPPMHVWECSIRJ-MDZDMXLPSA-N

Analysis Note

Acid value196-204Congealing temperature10°C maxDescriptionTo pass testIodine value85-95Mineral acidsTo pass testNeutral fat or mineral oilTo pass testResidue after ignition0.01% maxSolubilityTo pass test

Legal Information

OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service