Skip to Content
MilliporeSigma
All Photos(1)

Documents

NC010

Sigma-Aldrich

Nerve Growth Factor 7.0s

Synonym(s):

holo-NGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160405
NACRES:
NA.41

biological source

mouse

Assay

97% (SDS-PAGE)

form

solid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

Gene Information

mouse ... KLK4(56640)

General description

In the mouse submaxillary gland the neurotrophically active NGF, beta-NGF (2.5S), is part of a larger protein complex, 7S-NGF, which has the stoichiometry alpha2betagamma2 (+ 1 or 2 zinc ions) where the alpha and gamma subunits are members of the glandular kallikrein family of serine proteases, each of about 26 000 kDa molecular weight. The alpha and gamma subunits actually inhibit beta-NGF action and must be dissociated for biological activity to be observed.



This 7S complex has not been shown to occur in other mouse tissues, or in other species, probably because the alpha and gamma subunits, which are members of a larger glandular kallikrein family, apparently have a very restricted expression. The alpha and gamma subunits, members of the serine proteinase family, have quite similar amino acid sequences, although only the gamma subunit is catalytically active. The alpha subunit has undergone several mutations that preclude this function, including loss of the cleavage site needed for processing from the zymogen to the active form.
MOLECULAR WT.:

130-140 kD. 7S NGF is composed of 3 subunits; 1 beta subunit (2.5S), 2 alpha subunits and 2 gamma subunits.
Product Source: Mouse submaxillary gland (Darling & Shooter, 1984).

Application

The recommended concentration to be used in vitro for maintenance of sympathetic and immature sensory nerve cells is 50-100 ng/mL medium (Varon et al., 1967).

Physical form

Lyophilized. NC010 should be reconstituted with sterile DMEM at a concentration of

10 μg/mL and stored at -70°C. Medium with mNGF cannot be reused due to mNGF degradation.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Specific Activity: The neurotrophic activity is in the range of 0.5-50 ng/mL using rat PC12 cells cultured for 7-14 days

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The isolation of the mouse nerve growth factor protein in a high molecular weight form.
S Varon et al.
Biochemistry, 6(7), 2202-2209 (1967-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service