NA18/84
NovAseptic Valve, Spring Closed Valve Seal (for PTFE Diaphragm only)
All Photos(1)
About This Item
Recommended Products
material
valve (Spring Closed Valve Seal)
parameter
-1 to -6 bar pressure
0 to 140 °C temp. range
weight
0.2 kg0.44 lb
Application
The applied valve body, diaphragm and actuator may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Spring Closed Valve Seal is packaged in a closed box.