Key Documents
NA18/25
Millipore
Diaphragm for NOVASEPTIC® Valve
PTFE
Select a Size
1 PKG
Inquire
This item is not able to be purchased as a guest user. For further information, please contact us at +1 866 227 3811or
.
Select a Size
Change View
1 PKG
Inquire
About This Item
UNSPSC Code:
31411800
This item is not able to be purchased as a guest user. For further information, please contact us at +1 866 227 3811or
.
material
PTFE
PTFE device (polytetrafluoroethylene)
Quality Level
feature
hydrophobic (Surface)
parameter
-40-170 °C temp. range (Continous use)
-40-250 °C temp. range (Short term use)
weight
0.02 kg (0.044 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany