Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NA18

Sigma-Aldrich

Anti-Replication Protein A (Ab-2) Mouse mAb (RPA34-19)

This Anti-Replication Protein A (Ab-2) Mouse mAb (RPA34-19) is validated for use in Immunoblotting IF IP Paraffin Sections for the detection of Replication Protein A (Ab-2).

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$581.00

$581.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$581.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$581.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service