MZHAWG251
Microfil® with EZ-Pak®
ready-to-use, mixed cellulose esters (MCE) filter, pore size 0.45 μm, filter diam. 47 mm, sample volume -250 mL
Synonym(s):
Membrane filtration system
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE) filter
polypropylene
white filter
Agency
ISO 7704
sterility
sterile; ethylene oxide treated
feature
easily wetted with water
hydrophilic
ready-to-use
manufacturer/tradename
Microfil® with EZ-Pak®
parameter
-250 mL sample volume
filter diam.
47 mm
matrix
MF-Millipore™
pore size
0.45 μm pore size
input
sample type cosmetics
sample type wine
sample type sport drink
sample type beer
sample type cider
sample type beverage(s)
sample type food(s)
sample type bottled water
sample(s) (environmental)
sample type soft drink
suitability
suitable for microbiology
application(s)
bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc
compatibility
for use with EZ-Fit® Filtration Manifold
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Convenient and easy to use
- Sterilized and ready-to-use device- saves time and expense
- Reduce laboratory workload
- Faster filtration rate due to larger surface area
Principle
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service