Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MXSMBPA48

Millipore

Milliflex Cassette prefilled with Baird Parker Agar

Baird Parker Agar, for Staphylococcus spp., pack of 48 tests, suitable for bioburden testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160801
NACRES:
NA.74

form

plate (pre-poured agar)

shelf life

6 mo.

feature

ready-to-use

packaging

cassette of 10 mL
pack of 48 tests

manufacturer/tradename

Milliflex®

color

opalescent yellow-brown

input

sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water

pH

6.8±0.2 (25.0 °C)

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex®

shipped in

dry ice

storage temp.

2-8°C

suitability

Staphylococcus spp.

Culture Medium

Baird Parker

Analysis Note

QC organisms
S. aureus (ATCC 6538)

E. coli (ATCC 25922)

P. mirabilis (ATCC 12453)
Target Microorganisms: Staphylococcus species
Typical S. aureus colonies are black. If other microorganisms grow, colonies are white to brown

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service