Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MXRPARA06

Millipore

Milliflex® Rapid Reagent

pkg of 6 vials

Synonym(s):

Rapid testing reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NB.79

form

liquid

Quality Level

packaging

pkg of 6 vials

manufacturer/tradename

Milliflex®

dilution

(Environmental analysis)

application(s)

bioburden testing
environmental
food and beverages
pharmaceutical
sterility testing

compatibility

Milliflex® Rapid

storage temp.

2-8°C

General description

Milliflex® Rapid Reagent is a part of Milliflex® Rapid Microbiology Detection and Enumeration system. The reagents are sprayed onto the membrane using the Milliflex® Rapid AutoSpray Station.

Application

Milliflex® Rapid Reagents are used to spray onto the membrane to perform a final read out on the detection tower to check for contaminations.

Components

  • 6 x ATP releasing reagent vials (5 ml per bottle)
  • 6 x Luer-lok® caps
  • 6 x Vials adapters

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service