Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MXQULTVP1

Millipore

Milliflex Quantum Validation Protocol

The Milliflex Quantum validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
25169090
NACRES:
JA.11

Quality Level

manufacturer/tradename

Milliflex® Quantum

input

sample type pharmaceutical(s)

application(s)

industrial qc

shipped in

ambient

General description

Service Type: Validation Services

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service