Key Documents
MXPPA4RP1
Millipore
Milliflex® Plus Essential Requalification Protocol
A4 format
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
Pricing and availability is not currently available.
compatibility
for use with Milliflex® Plus pump.
General description
Milliflex® Plus Essential Requalification protocol is designed as a ready to use guide to perform the requalification of the Milliflex® Plus pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals.
Application
Milliflex® Plus Essential Requalification protocol is used for the re-qualification of Milliflex® Plus pump which is used to perform bioburden testing for pharmaceutical products.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany