Skip to Content
MilliporeSigma

MTRACEBD1

M-TRACE® Back Up Drive

for use with M-TRACE® All-in-One Computer

Synonym(s):

External Hard disk

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
JA.12
UNSPSC Code:
43210000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of -1 unit

application(s)

pharmaceutical
sterility testing

compatibility

Steritest® Symbio pump, for use with M-TRACE® All-in-One Computer

General description

The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for sterility testing and other regulated processes, ensuring trusted and efficient data recording. Click here to get more details on the overall M-Trace® solution.

The M-TRACE® Back Up Drive is a recommended external memory storage device used for the safe data backup recorded by the M-TRACE® All-in-One Computer (MTRACEPC1). It has a storage capacity of up to 512 GB. The M-TRACE® All-in-One Computer is the central control unit to create or start test records. All data sent from the connected devices, such as the Steritest® Symbio pump, will be collected and stored on the computer. These data can be backed up and stored in the M-TRACE® Back Up Drive

Application

The M-TRACE® Back Up Drive is a recommended external memory storage device used for safe backup of data recorded by the M-TRACE® All-in-One Computer that is used for sterility testing and other quality control workflows

Other Notes

Data storage capacity is 512 GB

Legal Information

M-TRACE is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library