Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MSPHEAD01

Millipore

Micropresure® opener

Milliflex® Plus Pump, suitable for bioburden testing, water monitoring

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,630.00

$1,630.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
$1,630.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$1,630.00


Please contact Customer Service for Availability

Request more information

material

316L stainless steel

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex®

input

sample type food(s)
sample type beverage(s)

application(s)

bioburden testing
water monitoring

clinical
environmental
food and beverages
industrial qc

compatibility

Milliflex® Plus Pump

shipped in

ambient

General description

Micropresure® opener head is a pump head adapter for Micropresure® On-Line Filtration Samplers. By reducing the sampling process to a few simple steps performed within a closed system, the Micropresure® On-Line Filtration Sampler improves micro- biological testing of purified water at the point-of-use. MSOpener head is compatible with the Milliflex® Plus pump. The standard head in the Milliflex® Plus pump can be lifted off to replace it with the MSOpener head.

Application

MSOpener head is an accessory of the Milliflex® Plus Pump used during filtration of the test samples for, bioburden testing, pharmaceutical product analysis, and water monitoring.

Preparation Note

Sterilization Method
Autoclavable

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service