Skip to Content
MilliporeSigma
All Photos(1)

Documents

MSNANLY

Millipore

Multiscreen HTS Filter Plate, Styrene

Clearing of bacterial cell lysates

Synonym(s):

Montage Plasmid Miniprep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.15

material

polystyrene

Quality Level

sterility

non-sterile

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

DNA purification: suitable

plate size

96 wells

General description

MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research. MultiScreen HTS plates are specifically developed for high throughput use with automated workstations. Plasmid purification is a widely employed method in molecular cloning, transformation, and recombinant protein expression. Plasmid purification kits are designed to accommodate different bacterial culture sizes and the corresponding plasmid yield.

Application

Multiscreen HTS Filter Plate, Styrene is suitable for use in the clearing of bacterial lysates with the MultiScreen HTS vacuum manifold prior to plasmid purification using MSNUPSD50.

Features and Benefits

  • Enhanced handling is achieved with rigid sidewalls.
  • Abundant surface area for barcode labels.
  • Prevents any cross-contamination during incubation.
  • High throughput screening

Physical form

Color Code: Clear

Other Notes

Automation Compatible: Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service