Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MMVHPKIT01

Milliflex Oasis® VHP resistant Vacuum Pump - Kit

for use with Milliflex Oasis® systems, suitable for bioburden testing, sterility testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.74

Agency

EP 2.6.1
USP 71

product line

Milliflex Oasis®

packaging

pkg of 1 kit

technique(s)

Rapid Sterility Testing: suitable

application(s)

bioburden testing
sterility testing

pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® systems

shipped in

ambient

General description

Milliflex Oasis® VHP resistant filtration Kit is a complete membrane filtration kit comprised of the pump and associated accessories for cleaning and maintenance.

The Milliflex Oasis® VHP resistant pump is a membrane filtration pump, and the filtration heads are vaporous hydrogen peroxide (VHP) resistant. This pump is equipped to be used inside isolators.

The detailed contents of the Kit are mentioned in “Components” section. Scroll below to find!

Application

Milliflex Oasis® VHPresistant filtration Kit is a perfect membrane filtration solution used forbioburden and sterility testing in isolators The system is compatible with awide range of filterable samples from pharmaceutical, beverage and personalcare industries.

Features and Benefits

  • Streamlines filtration steps
  • Compatible with the isolator environment and VHP decontamination
  • Compatible with the Milliflex® Rapid System 2.0 to perform rapid sterility testing in a clean environment (isolator)

Components

Milliflex Oasis® VHP resistant filtration Kit iscomprised of:
  • 2 x Milliflex Oasis® VHP pumps - MMPPLUVHP
  • Milliflex Oasis® electrical cable for connectingtwo Oasis VHP pumps - MMCABLEVH
  • Milliflex Oasis® Pump filtration head -MMHEADMM1
  • Sanitization kit for Milliflex Oasis® VHPresistant filtration pump - MMSANKITVHP
  • Stainless steel cover for Sanitization kit of Milliflex Oasis® VHP pump - MMSANKITCVR
  • Stainless steel plug for Milliflex Oasis® VHP pump outlet -MMVHPPLUG
  • Milliflex Oasis® T connectors for tubings for connecting twopumps - MMTCNNECT
  • Milliflex® Quantum membrane removal tool - REMRACKMM
  • Milliflex Oasis® vacuum gauge for pump performance testing- MMGAUGEMM
  • Power Supply- not included (sold Separately region specific under VHPPWRSP--)

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service