Documents
MMSANKITVHP
Millipore
Milliflex Oasis® VHP resistant pump, internal flow sanitization kit
for use with Milliflex Oasis® VHP resistant pump
About This Item
UNSPSC Code:
41122409
NACRES:
NB.75
material
stainless steel
Sterilization Compatibility
autoclavable compatible
packaging
pkg of 1 kit
manufacturer/tradename
Milliflex®
technique(s)
Rapid Sterility Testing: suitable
application(s)
bioburden testing
pharmaceutical
sterility testing
compatibility
for use with Milliflex Oasis® VHP resistant pump
General description
Milliflex Oasis® system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The Milliflex Oasis® VHP resistant pump is a new generation of vacuum pump for filtration in VHP isolator environments. The Milliflex Oasis® VHP resistant pump, internal flow sanitization kit is an accessory to be used to decontaminate the interior of the Milliflex Oasis® VHP resistant pump.
Application
The Milliflex Oasis® system finds its application in bioburden testing and water monitoring of Pharmaceutical Products Analysis
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Articles
Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.