Skip to Content
MilliporeSigma
All Photos(1)

Documents

MM2PA4FT1

Millipore

Milliflex Oasis® Pump Functional Test Report

Verification Report for Core Functions in A4 Format

Synonym(s):

System Verification Report

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
ND.05

manufacturer/tradename

Milliflex Oasis®

compatibility

for use with Milliflex Oasis® Vacuum Pump

General description

Milliflex Oasis® Pump Functional Test Report is designed as a light guide for users to perform a verification exercise of the core functions of the equipment after purchase and before starting with routine filtration of samples. Milliflex Oasis® Pump is used to carry out water and bioburden testing through filtration process using EZ-Fit® filtration units.
The verification protocol in A4 format ensures optimal performance of the Milliflex Oasis® Pump.

Application

Milliflex Oasis® Pump Functional Test Report in A4 format is used to perform verification of core functions of the equipment.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service