Skip to Content
MilliporeSigma
All Photos(2)

Documents

MISELLABSE

Millipore

MilliSentials Lab Labeling System

Europe

Synonym(s):

Laboratory labeling system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
44102408
NACRES:
IA.21

Quality Level

General description

Other Notes:

For existing users of ZD620 and ZD621 printers: Download the updated “MilliSentials” Lab Labeling System Software (with barcode printing features) here.
The MilliSentials Lab Labeling System provides a complete laboratory labeling solution with laboratory grade labels, a compact WIFI capable printer, and custom-designed laboratory labeling software. The laboratory labels are capable of withstanding temperatures from -196°C (liquid nitrogen) to 100°C (boiling water) and provide resistance to harsh chemical solvents (including ethanol, DMSO, xylene). The labels have also been designed with perforations allowing one size to fit across a wide range of laboratory vessels including microcentrifuge tubes, cryovials, petri dishes, cell culture plates, and reagent bottles. The custom-built software features pre-constructed label templates for laboratory vessels, allows labels to be saved for quick repeat printing, and features a spreadsheet upload function for large batch printing. The MilliSentials Lab Labeling System offers a complete labeling system specifically designed for the laboratory.

Properties-
1. MilliSentials Lab Printer-
Width:202 mm (8.0 in)
Height:192 mm (7.5 in)
Weight:2.5 kg (5.5 lbs)
Connection / Communication: USB, F802.11ac WIFI, Bluethooth 4.1
Resolution:300 dpi
Power:Auto-detectable (PFC compliant), 100-240 VAC, 50-60Hz
Warranty: 3 year

2. MilliSential Lab Adaptable Labels-
Label: MilliSentials Lab Adaptable Labels
Vessel Compatibility:0.5 mL Tubes, 0.6 mL Tubes, HPLC vials, 1 mL Tubes, 2 mL Tubes, Cryovials, Plates, Petri Dishes, 15 mL Conical Tubes, 50 mL Conical Tubes, Any Vessel Larger than 5 mL
Temperature Range:-196°C to 100°C
Chemical Compatibility: Water, Isopropyl Alcohol (≤95%), Ethanol (≤95%), Dimethyl - sulfoxide (≤70%), Methanol (≤95%), Acetonitrile (≤75%), Bleach (≤70%), Xylene (≤95%), Acetone (≤95%)

3. MilliSentials Lab Printer Ribbon-
Ribbon Capacity: ~6400 labels printed from 1 roll of ribbon

4. MilliSentials Lab Labeling System Software - USB Key
Software: MilliSentials Lab Labeling System Software
Software Compatibility: Microsoft Windows 10 OS, version 1909 OS Build 18363.1016 Macintosh(Mac) macOS Catalina, version 10.15.6

Application

Laboratory Sample Labeling

Components

1. MilliSentials Lab Printer
2. MilliSential Lab Adaptable Labels -1 roll
3. MilliSentials Lab Printer Ribbon - 1 roll
4. MilliSentials Lab Labeling System Software - USB Key

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Streamline sample, reagent, and chemical identification using the MilliSentials™ Lab Labeling System - impervious, adaptable labels tailored for research labs.

Streamlined, ergonomic, essential lab equipment solutions for handling, labeling, pipetting, aliquoting, and homogenizing samples in research laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service