Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MHA000P2P

Millipore

Pseudomonas Selective Broth

Ampoule, Selective media for Pseudomonas. Inhibits other bacteria.

Synonym(s):

PS broth

Sign Into View Organizational & Contract Pricing

Select a Size

50
$144.00

$144.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
50
$144.00

About This Item

UNSPSC Code:
41106225
eCl@ss:
32160801
NACRES:
NA.74

$144.00


Please contact Customer Service for Availability

Request more information

Quality Level

sterility

sterile

form

liquid

shelf life

12 mo.

feature

ready-to-use

packaging

pack of 50 tests

capacity

2 mL

filling volume

2 mL

color

amber to green-amber, some particulates may be present in final product

input

sample type beer
sample type soft drink
sample type bottled water
sample type sport drink
sample type food(s)
sample type beverage(s)
sample type cosmetics
sample type wine
sample type cider

pH

7.1±0.2 (25.0 °C)

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc (microbial monitoring)
microbiology
water monitoring

compatibility

for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®

shipped in

wet ice

storage temp.

2-8°C

suitability

Pseudomonas spp.

General description

Pseudomonas Selective Broth is a Selective media for the growth of Pseudomonas species and inhibits the growth of other bacteria. Tryptone and gelatin peptone in the media supplies nitrogenous and carbonaceous compounds, long chain amino acids, and other essential growth nutrients.{13} This medium is recommended for enumeration of Pseudomonas species from meat and meat products. It can also be used for clinical samples. Pseudomonas species produce small colourless to translucent yellow colonies when incubated at 25−35 °C (35 °C during 24h is favourable to P. aeruginosa, 25 °C for Pseudomonas species)

Application

Pseudomonas Selective Broth is used to enumerate Pseudomonas species in samples such as Beer, Bottled Water, Cider, Soft Drink, Sport Drink and Wine.(2) This media finds its application in Clinical lab, Cosmetics, Environmental analysis, Food and Beverage, Industrial QC, Microbial monitoring and Water Monitoring

Packaging

2 mL plastic ampoules

Specifications

Incubation Temperature: 25–35 °C (35 °C during 24h is favorable to P. aeruginosa, 25 °C for Pseudomonas species)
Incubation Time: 24–72 hrs

Culture Medium

Pseudomonas Selective

Analysis Note

Pseudomonas species produce small colorless to translucent yellow colonies
QC organisms
P. aeruginosa (ATCC 27853)
Target Microorganisms: Pseudomonas species

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manual of clinical microbiology
Jorgensen JH et al.
Manual of Clinical Microbiology, 2 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service