MB1HC23CL3
Millistak+® Depth Filter in μPod format
B1HC, filtration area 23 cm², Process Development Scale
Synonym(s):
Millistak+Depth Filter in Pod format, B1HC media series, 23 cm2 surface area, Luer fitting connection
Select a Size
$317.00
Select a Size
About This Item
$317.00
Recommended Products
Product Name
Millistak+® Depth Filter in μPod format, B1HC, cellulose fiber, filtration area 0.0023 m2, hydrophilic
material
cellulose fiber
filter aid (inorganic)
glass fiber-reinforced polypropylene
Quality Level
description
Post-centrifuge or settled permeate containing cellular particulate
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
1.0 bar max. differential pressure (15 psid) at 40 °C (Reverse)
2.1 bar max. differential pressure (30 psid) at 40 °C (Forward)
H
6.6 cm (2.6 in.)
L
8.9 cm (3.5 in.)
filtration area
0.0023 m2
impurities
<0.25 EU/mL (LAL test, Aqueous extraction)
gravimetric extractables
420-750 mg/ft2
input
sample type liquid
conductivity
3.64 – 10.5 μS/cm
filter grade
B1HC
shipped in
ambient
General description
This B1HC media series delivers optimal performance through a gradient density matri, ideal for secondary clarification post tangential flow filtration, or depth filtration or centrifugation applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Application
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Millistak+® HC dual-action media improves prefiltration and compresses clarification
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Preparation Note
2 autoclave cycles of 60 minutes at 123 °C.
Analysis Note
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service