Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAVFLTTR1

Millipore

MAS-100 VF® Portable Air Sampler Test Records Protocol

for use with MAS-100 VF® Microbial Air Samplers

Synonym(s):

System Verification Report

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
JA.11
Pricing and availability is not currently available.

availability

not available in North America

compatibility

for use with MAS-100 VF® Microbial Air Samplers

Looking for similar products? Visit Product Comparison Guide

General description

MAS-100 VF® Pump Functional Test Record Protocol(Letter Format) contains test descriptions and test results sheets to perform the IQ and OQ of one air sampler unit. It must be ordered together with the Validation Protocol MAVFLTVP2. The protocol follows GMP, EP, USP and JP regulatory guidelines. Only available in North America.

Application

MAS-100 VF® Validation Protocol is used to validate microbial air sampling system with ease and consistent performance while following regulatory guidelines.

Legal Information

MAS-100 VF is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service