Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MATAHCL00

Millipore

MultiScreen Sealing Tape, clear, non-sterile

Tape used for sealing top of wells when running partial plates or sealing bottom of harvest plates for coincidence counting.

Synonym(s):

Clear sealing tape

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$117.00

$117.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
$117.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

$117.00


Please contact Customer Service for Availability

Quality Level

sterility

non-sterile

product line

MultiScreen®

manufacturer/tradename

MultiScreen®

shipped in

ambient

General description

No matter what application you’re working with, Millipore has a plate developed to meet your specific needs. MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.



HTS Filter Plates (Plate catalogue numbers that start with MS)

- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.



- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.



Classic Filter Plates (Plate catalogue numbers that start with MA)

- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.



Accessory – MultiScreenHTS Vacuum Manifold

- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.



Millicell Assay Plates

- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service