Skip to Content
MilliporeSigma

Skip To

MAC123

Anti-HSV-Tag® Antibody, clone HSV 78

clone HSV 78, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
$637.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
HSV 78, monoclonal
Technique(s):
western blot: suitable
Application:
WB
Citations:
1

$637.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified immunoglobulin

clone

HSV 78, monoclonal

species reactivity (predicted by homology)

all

technique(s)

western blot: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

The HSV•Tag is a mouse monoclonal antibody that has high specificity and affinity for an 11-amino acid peptide derived from Herpes Simplex Virus (HSV) glycoprotein D. The peptide is encoded by the HSV•Tag sequence in appropriate expression vectors and by the Positive Control 36mer in the Novagen NovaTope System.
HSV•Tag and NovaTope are registered trademarks of Merck KGaA, Darmstadt, Germany.
~45 kDa observed. An uncharacterized band may be observed at ~40 kDa, when using the Posi-Tag Epitope Tag Control protein. (Note: The Posi•Tag Epitope Tag Control which contains an HSV•Tag sequence, is observed at ~45 kDa)

Immunogen

Linear peptide corresponding to the HSV•Tag sequence.

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Epitope Tags
This HSV•Tag antibody is validated for use in WB for the detection of the HSV•Tag protein.

Biochem/physiol Actions

Predicted to react with proteins containing the HSV•Tag.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Posi-Tag Epitope Tag Control protein containing the HSV•Tag sequence.
Evaluated by Western Blotting in Posi-Tag Epitope Tag Control protein containing the HSV•Tag sequence.

Western Blotting Analysis: 0.001 µg/mL of this antibody detected HSV•Tag in 50 ng of Posi-Tag Epitope Tag Control protein containg the HSV•Tag sequence.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 69171-4; 69171-3

Legal Information

HSV-TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200695C7706MABF2820
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified antibody

clone

HSV 78, monoclonal

clone

P5D4, monoclonal

clone

P5D4, monoclonal

clone

AP33, monoclonal

isotype

IgG1κ

isotype

IgG1

isotype

IgG1

isotype

IgG1κ

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

shipped in

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters