Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX8415

Sigma-Aldrich

Anti-COVID-19 & SARS-CoV, Nucleoprotein Antibody, 1055

suitable for ELISA, suitable for western blot

Synonym(s):

SARS-CoV-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1055, monoclonal

storage condition

avoid repeated freeze/thaw cycles

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

General description

This mouse monoclonal Anti-COVID-19 & SARS-CoV, Nucleoprotein, 1055, Cat. No. MABX8415 is tested for use in ELISA and Western Blotting, for the detection of COVID-19 Nucleoprotein.

Specificity

COVID-19 & SARS Coronavirus Nucleoprotein (NP)

Immunogen

Recombinant SARS-CoV (aa 1-49)

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus.

Physical form

Purified in10mM Phosphate Buffered Saline with 0.1% sodium azide, pH 7.0.

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Short Term: 2-8°C. Long Term: -20°C. Avoid repeated freezing and thawing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service