Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX45

Sigma-Aldrich

Anti-Dengue Virus (DENV-pan NS1)

Synonym(s):

Anti-DENV-pan NS1 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX45-1KC)
pkg of 10 mg (MABX45-10KC)

technique(s)

ELISA: suitable
immunodiffusion: suitable
immunofluorescence: suitable

isotype

IgG1

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

NS1 protein of all four Dengue serotypes

Immunogen

Partially purified Native Dengue Virus

Application

This mouse monoclonal Anti-Dengue Virus (DENV-pan NS1), Cat. No. MABX45, is tested for use in Immunofluorescence, ELISA and Lateral Flow Assay, for the detection of NS1 protein of all 4 serotypes.

Target description

Dengue virus is an arthropod-borne virus (arbovirus). Dengue fever is caused by any of four related flaviviruses spread to people through the bite of infected Aedes species mosquitoes. This antibody recognizes a common epitope on the NS1 protein in all four Dengue strains.

Linkage

Recommended matched pair MABX45 capture and MABX44 detection.

Physical form

Purified mouse monoclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.2 with 0.1% sodium azide.

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service