Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX4187

Sigma-Aldrich

Anti- human myeloperoxidase (MPO) clone 187

clone 187, from mouse

Synonym(s):

Myeloperoxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

187, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

Gene Information

human ... MPO(4353)

General description

Antibody recognizes Myeloperoxidase. Monitoring of level of Myeloperoxidase (MPO) in plasma can be useful to assist in the diagnosis critical cardiac conditions such as a Myocardial Infarction and also as a prognostic marker for other Cardiovascular Diseases (CVD s)

Application

Confirmed matched pairing. Capture Antibody MABX4187-1KC; Detector Antibody MABX4043-1KC
Research Category
Cell Structure
This mouse monoclonal Anti- human myeloperoxidase (MPO) clone 187 Antibody, Cat. No. MABX4187-1KC is validated for use in and Western Blotting and ELISA, for the detection of MPO.

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing PBS (pH 7.4) with 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service