Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX4144

Sigma-Aldrich

Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 144

clone 144, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

144, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

Gene Information

human ... IGFBP4(3487)

General description

Antibody recognizes the Human insulin like growth factor binding protein 4 (IGFBP-4). IGFBP-4 is a member of the Insulin like growth factor family and is responsible for binding insulin like growth factors I and II. Research has linked levels of C-term (CT-IGFBP-4) and N-term (NT-IGFBP-4) to certain cardiac events, offering potential as a novel cardiac biomarker

Specificity

Human

Application

Confirmed matched pairing. Capture Antibody MABX4013-1KC or Capture Antibody MABX4182-1KC; Detector Antibody MABX4144-1KC
Detect IGFBP-4 using this mouse monoclonal , Cat. No. MABX4144-1KC, validated for use in and Western Blotting and ELISA.
Research Category
Signaling

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing PBS (pH 7.4) with 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service