Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX30

Sigma-Aldrich

Anti-Zika Virus (ZIKV-NS1)

Synonym(s):

Anti-ZIKV-NS1 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX30-1KC)
pkg of 10 mg (MABX30-10KC)

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

No reactivity with Dengue Virus or Chikungunya Virus

Immunogen

Recombinant Zika Virus NS1 Protein

Application

This mouse monoclonal antibody, Cat. No. MABX30, is tested for use in Lateral Flow Assay and Enzyme Immunoassay (ELISA), for the detection of Recombinant Zika Virus NS1 Protein.

Target description

Zika virus (ZIKV) is a member of the virus family Flaviviridae. Zika is spread mostly by the bite of infected Aedes species mosquitoes, such as Ae. aegypti and Ae. albopictus. Its name comes from the Ziika Forest of Uganda, where the virus was first isolated in 1947.  Zika can be passed from a pregnant woman to her fetus. Infection during pregnancy can cause certain birth defects.

Linkage

Recommended Matched Pair MABX30 and MABX31

Physical form

Purified mouse monoclonal in buffer containing 0.015 M Potassium Phosphate, pH 7.2 with 0.85% Sodium Chloride and 0.05% Sodium Azide

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service