Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT887

Sigma-Aldrich

Anti-PL Scramblase 1 Antibody, clone 9A7

clone 9A7, from mouse

Synonym(s):

Phospholipid scramblase 1, PL scramblase 1, Ca(2+)-dependent phospholipid scramblase 1, Transplantability-associated protein 1, NOR1, TRA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9A7, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Plscr1(22038)

General description

Phospholipid scramblase 1 (UniProt Q9JJ00; also known as Ca(2+)-dependent phospholipid scramblase 1, Nor1, PL scramblase 1, Tra1, Transplantability-associated protein 1) is encoded by the Plscr1 (also known as Tra1b, Tras1) gene (Gene ID 22038) in murine species. Plasma membrane phospholipids are distributed asymmetrically between the inner and outer leaflets. Such asymmetrical distribution collapses in response to blood coagulation and apoptosis, resulting in phospholipid “scrambling” between the two leaflets of the plasma membrane. Flippases, floppases, and scramblases are three types enzymes known to mediate transbilayer lipid motion. Flippases and floppases function via ATP-dependent mechanism, while scramblases mediate transbilayer movement in a non-selective and energy-independent manner. Originally identified in 1996 as a 37 kDa erythrocyte type II transmembrane protein that mediates calcium-dependent membrane phospholipids redistribution, PL Scramblase 1 is the protein product encoded by the founding member of the PLSCR family of genes (PLSCR1-5). All PLSCR family members, with the exception of PLSCR2, possess a proline-rich N-terminal region containing PxxP and PPxY domains, a cysteine-rich region, a conserved calcium-binding domain (EF-hand-like), and a putative transmembrane region enriched in hydrophobic amino acids. In addition, PLSCR1 contains a nuclear localization signal (NLS) and a DNA-binding domain that are essential for its nuclear localization and associated nuclear function. In addition to PLSCRs, TMEM16 and XKR family members have also been reported to mediate scramblase activity.

Specificity

Clone 9A7 reacts with murine PL Scramblase 1, and does not cross-reacts with murine PL Scramblase 2/3/4 or human PL Scramblase 1.

Immunogen

Epitope: Cytoplasmic domain
Recombinant protein corresponding to the cytoplasmic domain of mouse PL Scramblase 1.

Application

Anti-PL Scramblase 1 Antibody, clone 9A7 is an antibody against PL Scramblase 1 Antibody for use in Western Blotting.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Western Blotting Analysis: A representative lot detected PL Scramblase 1 in mouse lung homogenates. Clone 9A7 also detected LPS treatment-induced PL Scramblase 1 upregulation in liver using total tissue homogenates, as well as cytosolic and nuclear extracts from LPS-treated and untreated mice (Lu, B., et al. (2007). Biochim. Biophys. Acta. 1771(9):1177-1185).

Quality

Evaluated by Western Blotting in mouse thymus tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected PL Scramblase 1 in 10 µg of mouse thymus tissue lysate.

Target description

~38 kDa observed. Two additional bands shown correspond to heavy and light chains of endogenous IgG from the mouse thymus tissue.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service