Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT538-AF488

Sigma-Aldrich

Anti-Lamin A/C Antibody, clone 2E8.2 Antibody, Alexa Fluor 488 Conjugate

clone 2E8.2, from mouse, ALEXA FLUOR 488

Synonym(s):

70 kDa lamin, Renal carcinoma antigen NY-REN-32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E8.2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Lamin-A/C (UniProt: P02545; also known as 70 kDa lamin, Renal carcinoma antigen NY-REN-32) is encoded by the LMNA (also known as LMN1) gene (gene ID: 4000) in human. Lamins are components of the nuclear lamina, a fibrous layer on the nucleoplasmic side of the inner nuclear membrane, which is thought to provide a framework for the nuclear envelope and may also interact with chromatin. Proteolytic cleavage of 18 residues from the C-terminal of prelamin-A/C results in the production of lamin-A/C. Proteolytic cleavage requires prior farnesylation and methylation, and absence of these blocks cleavage. The prelamin-A/C maturation pathway includes farnesylation of CAAX motif, ZMPSTE24/FACE1 mediated cleavage of the last three amino acids, methylation of the C-terminal cysteine and endoproteolytic removal of the last 15 C-terminal amino acids. Farnesylation of prelamin-A/C also facilitates nuclear envelope targeting. Lamin A and C are present in equal amounts in the lamina of mammals. Lamin plays an important role in nuclear assembly, chromatin organization, nuclear membrane and telomere dynamics. Lamin C differs from canonical form A in the following manner: amino acids 567-572 are changed from GSHCSS to VSGSRR and form C does not amino acids 573-664 seen in canonical form A.

Specificity

Clone clone 2E8.2 specifically recognizes Lamin A/C in HeLa cells. This antibody has been shown to bind to an epitope between amino acids 464-572 in the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 109 amino acids from the C-terminal half of human Lamin A/C.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Lamin A/C in A431 cells.

The unconjugated antibody (Cat. No. MABT538) is shown to be suitable also for Flow Cytometry, Immunohistochemistry, and Immunoprecipitation applications.
Research Category
Cell Structure
This mouse monoclonal Anti-Lamin A/C Antibody, clone 2E8.2, Alexa Fluor 488 Conjugate, Cat. No. MABT538-AF488 is used for the detection of Lamin A/C by Immunocytochemistry.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Lamin A/C in HeLa cells.

Target description

74.14 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Protein G purified
Purified mouse monoclonal antibody in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service