Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT524

Sigma-Aldrich

Anti-Cortactin (p80/85) Antibody, clone 5G3.2

clone 5G3.2, from mouse

Synonym(s):

Src substrate cortactin, Amplaxin, Oncogene EMS1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$680.00

$680.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1329

Request a Bulk Order

Select a Size

Change View
100 μL
$680.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$680.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1329

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5G3.2, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CTTN(2017)

General description

Src substrate cortactin (UniProt Q14247; also known as Amplaxin, Oncogene EMS1) is encoded by the CTTN (also known as EMS1) gene (Gene ID 2017) in human. Cortactin is a cytosolic protein that promotes polymerization and rearrangement of the actin cytoskeleton, especially the actin cortex around the cellular periphery. Cortactin is activated via phosphorylation by upstream kinases in response to extracellular stimuli. Src is known to phosphorylate cortactin on Tyr421, Tyr466, and Tyr482, leading to cortactin interaction with F-actin and Arp2/3. Erk can also phosphorylate cortactin on Ser405 and Ser418, causing recruitment of additional actin nucleation-promoting factors (NPFs), most importantly N-WASP, to the Arp2/3/ complex. Phosphorylated cortactin thus recruits and stabilizes Arp2/3 complex on preexisting F-actin, providing a nucleation site for a new actin branch to form. Cortactin overexpression in tumor cells enhances invasiveness as a result of highly active lamellipodia formation, also known as invadopodia. Cortactin contains an N-terminal aspartic acid-aspartic acid-tryptophan (DDW) sequence (a.a. 29-31 in chicken; a.a. 20-22 in human & mouse) that mediates its interaction with Arp2/3, followed by six and a half repeat sequences (a.a. 80-125, 126-162, 163-199, 200-236, 237-273, 274-310, 311-333 in chicken; a.a. 80-116, 117-153, 154-190, 191-227, 228-264, 265-301, 302-324 in human and mouse) that mediates F-actin interaction, a proline-rich coiled coil domain (a.a. 349-410 in chicken; a.a. 348-401 in human and mouse) that mediates Src SH3 domain interaction, and a C-terminal SH3 domain (a.a. 505-563 in chicken; a.a. 492-550 in human a.a. 488-546 in mouse) that contains Erk phosphorylation sites and is important for cortactin interaction with NPFs other than Arp2/3.

Specificity

Clone 5G3.2 epitope lies within the region containing cortactin repeats 3-6. Reactivity toward human spliced isoforms 2&3 that lack cortactin repeat 6 is possible, but has not been confirmed.

Immunogen

GST-tagged recombinant human cortactin fragment comprising cortactin repeats 3-6.

Application

Research Category
Cell Structure
This mouse monoclonal Anti-Cortactin (p80/85), clone 5G3.2 Antibody, Cat. No. MABT524 is validated for use in Western Blotting for the detection of Cortactin.

Quality

Evaluated by Western Blotting in 3T3/A31 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected cortactin in 10 µg of 3T3/A31 cell lysate.

Target description

~75 kDa observed. Target band size appears larger than the calculated molecular weights of 61.59/70.96/57.47 kDa (human isoform 1/2/3) and 61.25 kDa (mouse) due to posttranslational modification. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service