Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABT494

Sigma-Aldrich

Anti-LIMS1, clone 5G7 Antibody

ascites fluid, clone 5G7, from mouse

Synonym(s):

LIM and senescent cell antigen-like-containing domain protein 1, Particularly interesting new Cys-His protein 1, PINCH-1, Renal carcinoma antigen NY-REN-48

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$532.00

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$532.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5G7, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIMS1(3987)

General description

LIM and senescent cell antigen-like-containing domain protein 1 (LIMS1), also known as Particularly interesting new Cys-His protein 1 (PINCH-1; PINCH1; PINCH), or Renal carcinoma antigen NY-REN-48 is an adaptor protein which contains five LIM domains, or double zinc fingers. LIMS1 is likely involved in integrin signaling through its LIM domain-mediated interaction with integrin-linked kinase, found in focal adhesion plaques. It is also thought to act as a bridge linking integrin-linked kinase to NCK adaptor protein 2, which is involved in growth factor receptor kinase signaling pathways. Its localization to the periphery of spreading cells also suggests that Lims1 may play a role in integrin-mediated cell adhesion or spreading.

Immunogen

Purified recombinant fragment of human LIMS1 expressed in E. Coli

Application

ELISA: A representative lot detected control antigen (100 ng) and antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected LIMS1 in HeLa cells.

Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected LIMS1 in HepG2 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-LIMS1, clone 5G7 Antibody is validated for use in western blotting, ELISA, flow cytometry & immunofluorescence for the detection of LIMS1.

Quality

Evaluated by Western Blotting in A549, Jurkat and HeLa cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected LIMS1 in A549, Jurkat and HeLa cell lysates.

Target description

~37 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service