Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT196

Sigma-Aldrich

Anti-Endoplasmin GRP94 Antibody, clone 10C1.2

clone 10C1.2, from mouse

Synonym(s):

Endoplasmin, 94 kDa glucose-regulated protein, GRP-94, Heat shock protein 90 kDa beta member 1, Tumor rejection antigen 1, gp96 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$534.00

$534.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$534.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$534.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10C1.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HSP90B1(7184)

General description

94 kDa glucose-regulated protein (GRP94) is also called Endoplasmin, Heat shock protein 90 kDa beta member 1 (HSP90B1), Tumor rejection antigen 1 (TRA1) and gp96 homolog. GRP94 is involved in the processing and transport of secreted proteins. GRP94 also functions in endoplasmic reticulum associated degradation (ERAD), and has ATPase activity.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human Endoplasmin GRP94.

Application

Anti-Endoplasmin GRP94 Antibody, clone 10C1.2 is a highly specific mouse monoclonal antibody, that targets Glucose-Regulated Protein 94 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Endoplasmin GRP94 in human lung section tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in human lung tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Endoplasmin GRP94 in 10 µg of human lung tissue lysate.

Target description

~94 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service