Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT182

Sigma-Aldrich

Anti-Rab5A Antibody, clone 7D8.1

clone 7D8.1, from mouse

Synonym(s):

Ras-related protein Rab-5A, Rab5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RAB5A(5868)

General description

Rab5A is a membrane associated Ras related GTPase that is required for endosome fusion with the plasma membrane. Rab5A also contributes to filopodia extension and other cellular cytoplasmic extensions. Rab5A plays important roles in cellular differentiation, neurogenesis, exocytosis and protein transport. Rab proteins are a large family of GTP-binding proteins that regulate cellular membrane traffic and organelle identity. When membrane bound, each Rab has a very specific cellular location and recognizes distinct subsets of intracellular membranes, which allows Rab proteins to act as as key cellular regulators, determining the recruitment of downstream effectors to the correct membrane at the correct time. Recent research indicates that the localization of Rab proteins happens by sensing the stored curvature elastic energy in the membrane and the specific RabGEFs (Rab-activating guanosine diphosphate/guanosine triphosphate exchange factors) that associate with a particular Rab. Rab5A associates with Rabex-5 and this association provides the determinants for Rab5A specific membrane targets.

Immunogen

KLH-conjugated linear peptide corresponding to human Rab5A.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Rab5A in human testis tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Rab5A Antibody, clone 7D8.1 is validated for use in Western Blotting and Immunohistochemistry for the detection of Rab5A.

Quality

Evaluated by Western Blotting in HEK-293 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Rab5A in 10 µg of HEK-293 cell lysate.

Target description

~24 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service