Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MABT1577

Sigma-Aldrich

Anti-Mature Lamin A Antibody, clone 3E12

Synonym(s):

Mature Laminin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3E12, monoclonal

mol wt

calculated mol wt 74.14 kDa
observed mol wt ~70 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Prelamin-A/C (UniProt: P02545) is encoded by the LMNA (also known as LMN1) gene (Gene ID: 4000) in human. It is cleaved into Lamin-A/C (also known as 70 kDa Lamin, Renal carcinoma antigen NY-REN-32). Lamins are components of the nuclear lamina that provide a framework for the nuclear envelope and may also interact with chromatin. Lamin A and C are present in equal amounts in the lamina of mammals. They play an important role in nuclear assembly, chromatin organization, nuclear membrane, and telomere dynamics. Human lamin A and lamin C are identical in their first 566 amino acids with lamin C having six unique carboxyl-terminal amino acids. Lamin A is initially synthesized as prelamin A that undergoes several modifications in the carboxyl terminal region that allow incorporation of prelamin A into the nuclear envelope and its subsequent processing into the mature lamin A. Prelamin A contains a CAAX motif at its carboxyl-terminus, which signals a series of posttranslational enzymatic modifications, including farnesylation of the cysteine, cleavage of the AAX, and methylation of the carboxyl group of the newly exposed farnesylcysteine. Cleavage of 15 residues (aa 647-662) by ZMPSTE24 generates the final protein product. Unlike mature lamin A, prelamin A accumulates as discrete and localized foci at the nuclear periphery. Prelamin can accelerate smooth muscle cell senescence. It can act to disrupt mitosis and induce DNA damage in vascular smooth muscle cells, leading to mitotic failure, genomic instability, and premature senescence. Mutations in LMNA gene are known to cause Emery-Dreifuss muscular dystrophy that is characterized by weakness and atrophy of muscle without involvement of the nervous system. Some mutations have also been linked to familial type of lipodystrophy characterized by the loss of subcutaneous adipose tissue in the lower parts of the body. (Ref.: Casasola, A., et al. (2016). Nucleus 7(1); 84-102; Bonne, G., et al (1999). Nat. Genet. 21(3); 285-288).

Specificity

Clone 3E12 is a mouse monoclonal antibody that detects Lamin A. It targets an epitope within 6 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 6 amino acids from the C-terminal region of human mature Lamin A.

Application

Quality Control Testing

Evaluated by Western Blotting in HAP1 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected mature Lamin A in lysates from HAP1 cells, but not in lysates from HAP1 cells with ZMPSTE24 knockouts or LMNA knockouts.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected Mature Lamin A in wild-type HAP1 cells , but not in cells with ZMPSTE24 knockout. (Data courtesy of Dr. Stefan Schüchner, Dr. Egon Ogris, Max Perutz Labs, Medical University of Vienna, Austria).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Mature Lamin A in lysates from HAP1 cells. (Data courtesy of Dr. Stefan Schüchner and Dr. Egon Ogris, Max Perutz Labs, Medical University of Vienna, Austria).

Western Blotting Analysis: A representative lot detected Mature Lamin A in lysates from wild-type HAP1 cells, but not in cells with ZMPSTE24 or LMNA knockouts. (Data courtesy of Dr. Stefan Schüchner and Dr. Egon Ogris, Max Perutz Labs, Medical University of Vienna, Austria).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Mature Lamin A, clone 3E12, Cat. No. MABT1577, is a mouse monoclonal antibody that detects mature Lamin A and is tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service