Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1565

Sigma-Aldrich

Anti-Desmin Antibody, clone DE-U-10

Synonym(s):

CMD1I, CSM1, CSM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DE-U-10, monoclonal

mol wt

calculated mol wt 53.6 kDa
observed mol wt ~16 kDa

purified by

using protein G

species reactivity

hamster, human, rat, mouse

species reactivity (predicted by homology)

porcine

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

Porcine ... DES(396725)

Specificity

Clone DE-U-10 is a mouse monoclonal antibody that detects Desmin. It targets an epitope within 48 amino acids from the C-terminal half of the protein.

Immunogen

Purified full length porcine Desmin.

Application

Quality Control Testing

Evaluated by Western Blotting in mouse skeletal muscle tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Desmin in mouse skeletal muscle tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected Desmin in human skeletal muscle tissue extracts.

ELISA Analysis: A representative lot detected Desmin in ELISA applications (Debus, E., et al. (1983). EMBO J. 2(12):2305-12).

Western Blotting Analysis: A representative lot detected Desmin in Western Blotting applications (Debus, E., et al. (1983). EMBO J. 2(12):2305-12).

Immunofluorescence Analysis: A representative lot detected Desmin in Immunofluorescence applications (Debus, E., et al. (1983). EMBO J. 2(12):2305-12).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Desmin (UniProt: P02540) is encoded by the DES gene (Gene ID: 396725) in porcine. Desmin is a type III intermediate filament protein that is essential for maintaining the structural and functional integrity of muscle cells as part of the cytoskeletal network. It provides mechanical support to the muscle cells and facilitates the organization of contractile proteins within the cell. Additionally, it is involved in various cellular processes, including cell signaling, maintenance of cell shape, and intracellular transport. It is particularly abundant in cardiac, skeletal, and smooth muscle tissues. It is also enriched at postsynaptic membranes of neuromuscular junctions (NMJs). Mutations in the gene encoding Desmin have been associated with hereditary and sporadic myopathies, cardiomyopathies and myasthenic syndrome. (Ref.: Eiber, N., et al., (2020). Front. Mol. Neurosci. 13; Article 567084).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service