Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT1558

Sigma-Aldrich

Anti-Utrophin Antibody, clone MANCHO3 (8A4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MANCHO3 (8A4), monoclonal

mol wt

calculated mol wt 394.47 kDa
observed mol wt ~400 kDa

purified by

using protein G

species reactivity

human, mouse

species reactivity (predicted by homology)

canine

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... UTRN(7402)

General description

Utrophin (UniProt: P46939; also known as Dystrophin-related protein 1, DRP-1) is encoded by the UTRN (also known as DMDL, DRP1) gene (Gene ID: 7402) in human. Utrophin is a dystrophin related protein that displays a sequence homology with dystrophin and shares many of the protein-binding properties. It plays a role in anchoring the cytoskeleton to the plasma membrane. It contains 2 calponin homology domains in its N-terminal region (aa 31-135 and 150-255) and has 22 spectrin homology repeats. The C-terminal sequence in utrophin is highly conserved between dystrophin and utrophin in most species. Utrophin is distributed throughout the sarcolemma in fetal and regenerating muscle, but is shown to be down-regulated in normal adult muscle and is restricted to the myotendinous and neuromuscular junctions. Four different isoforms of utrophin have been described that are produced by alternative promoter usage and they differ in their distribution pattern. Isoform 1 displays high expression in muscle tissue. Similar to dystrophin, utrophin can also reported to form an extensive lateral association with actin filaments and protect actin filaments from depolymerization in vitro. However, utrophin binds laterally along actin filaments through contribution of acidic spectrin-like repeats rather than the cluster of basic repeats used by dystrophin. It has been suggested that utrophin could functionally compensate for the lack of dystrophin and studied have shown that transgenic overexpression of utrophin in dystrophin-deficient mdx mice results in full recovery for all known parameters of the dystrophic phenotype. (Ref.: Sengupta, K., et al. (2020). Sci. Rep. 10; 21492; Porter, JD., et al.(1998). J. Cell Sci. 111(13); 1801-1811; Rybakova, IN., et al., (2002). Mol. Biol. Cell. 13(5); 1512-1521).

Specificity

Clone MANCHO3 (8A4) is a mouse monoclonal antibody that detects Utrophin. It targets an epitope within 42 amino acids from the C-terminal region.

Immunogen

A recombinant fragment corresponding the 329 amino acids from the C-terminal region of human Utrophin.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Utrophin in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Utrophin in NIH3T3 cell lysate.

Immunofluorescence Analysis: A representative lot detected Utrophin in Immunofluorescence applications (McMorran, B.J., et al. (2016). Glycobiology. 26(10):1120-1132).

Immunohistochemistry Applications: A representative lot detected Utrophin in Immunohistochemistry applications (Nguyen T.M. et al. (1991). J Cell Biol. 115(6):1695-700).

Western Blotting Analysis: A representative lot detected Utrophin in Western Blotting applications (Morris, G.E., et al. (1998). Biochemistry. 37(31):11117-27; Nguyen T.M. et al. (1991). J Cell Biol. 115(6):1695-700; Lien, C.F., et al. (2012). J Biol Chem. 287(49):41374-85; Ghosh, T., et al. (2015). PLoS One;10(7):e0134809; Lindsay, A., et al. (2019). FEBS J.;286(13):2562-2576).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Utrophin, clone MANCHO3 (8A4), Cat. No. MABT1558, is a mouse monoclonal antibody that detects Utrophin and is tested for use in Immunofluorescence, , Immunohistochemistry, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service