Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1549

Sigma-Aldrich

Anti-MT5-MMP/MMP24 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3D2 also termed M24N 3D2, monoclonal

mol wt

calculated mol wt 70.46 kDa

species reactivity

mouse, human

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Mmp24(17391)

General description

Matrix metalloproteinase-24 (UniProt: Q9R0S2; also known as MMP-24, Matrix metalloproteinase-21, MMP-21, Membrane-type matrix metalloproteinase 5, MT-MMP 5, MTMMP5, Membrane-type-5 matrix metalloproteinase, MT5-MMP, MT5MMP) is encoded by the Mmp24 (also known as Mmp21, Mt5mmp) gene (Gene ID: 17391) in murine species. MMP-24 is a single-pass type I membrane protein that is mainly expressed in neuronal cells of both central and peripheral nervous systems. It mediates the cleavage of N-cadherin and acts as a regulator of neuro-immune interactions and neural stem cell quiescence. It is also involved in cell-cell interactions between nociceptive neurites and mast cells. MMP-24 is synthesized with a signal peptide (aa 1-41) and a propeptide (aa 42-128) that are subsequently cleaved off to generate functional MMP-24 (aa 129-618) or processed MMP-24 (aa 129-554). MMP-24 contains a cysteine switch motif (aa 110-117) that binds the catalytic zinc ion and inhibits its enzyme activity. The dissociation of the cysteine from the zinc ion upon the activation-peptide release results in the activation of enzyme.

Specificity

Clone 3D2 is a rat monoclonal antibody that detects MT5-MMP/MMP-24. It targets an epitope within 16 amino acids from the extracellular domain.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 16 amino acids from the extracellular domain of human MT5-MMP.

Application

Quality Control Testing

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2a.

Tested Applications


Western Blotting Analysis: A representative lot detected MT5-MMP/MMP24 in Western Blotting applications (Willem, M., et. al. (2015). Nature. 526(7573):443-7).

Western Blotting Analysis: A 1:200 dilution from a representative lot detected MT5-MMP/MMP24 in lysate from HEK293 cells over expressing MT5-MMP, but not in HEK293 cells over expressing MT1-MMP and in mock control HEK293 cell lysates (Negative). (Courtesy of Michael Willem, Biomedical Center, Ludwig Maximilian University of Munich, Germany).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MT5-MMP/MMP24, clone 3D2, Cat. No. MABT1549, is a rat monoclonal antibody that detects MT5-MMP and is tested for use in Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service