Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1538

Sigma-Aldrich

Anti-SNX9 Antibody, clone 6C6

clone 6C6, from rat

Synonym(s):

Sorting nexin-9, SH3 and PX domain-containing protein 1, Protein SDP1, SH3 and PX domain-containing protein 3A

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
100 μL
$624.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
100 μL
$624.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SNX9(51429)

General description

Sorting nexin-9 (UniProt: Q9Y5X1; also known as SH3 and PX domain-containing protein 1, Protein SDP1, SH3 and PX domain-containing protein 3A, SNX9) is encoded by the SNX9 (also known as SH3PX1, SH3PXD3A) gene (Gene ID: 51429) in human. SNX9 is widely expressed, with highest levels reported in heart and placenta, and lowest levels in thymus and peripheral blood leukocytes. SNX9 is localized at sites of endocytosis at the cell membrane and is detected on newly formed macropinosomes. It is transiently recruited to clathrin-coated pits at a late stage of clathrin-coated vesicle formation. SNX9 can exist as a homo dimer or homo oligomer and can also heterodimerize with SNX18. It displays four phosphatidylinositol 4,5-bisphosphate binding sites. It has a SH3 domain (aa 1-63), a SNX-type Phox homology (PX) domain (aa 250-361), and a BAR domain (aa 292-595). The PX domain mediates its interaction with membranes enriched in phosphatidylinositol phosphate. SNX9 can stimulate the GTPase activity of dynamin and both its SH3 domain and more C-terminal domains are required to efficiently stimulate this GTPase activity. Although it has high affinity for phosphatidylinositol 4,5-bisphosphate, but can also bind to membranes enriched in other phosphatidylinositol phosphates. SNX9 can be phosphorylated on tyrosine residues by TNK2 and phosphorylation promotes its activity in the degradation of EGFR. SNX9 is shown to be involved in endocytosis and intracellular vesicle trafficking, both during interphase and at the end of mitosis. It is required for efficient progress through mitosis and cytokinesis and also for normal formation of the cleavage furrow at the end of mitosis. (Ref.: Schobel, S., et al. (2008). J. Biol. Chem.283(21); 14257-14268).

Specificity

Clone 6C6 is a rat monoclonal antibody that detects human Sorting nexin 9 (SNX9). It targets an epitope with in the SH3 domain.

Immunogen

GST-tagged full-length recombinant human Sorting Nexin-9 protein.

Application

Anti-SNX9, clone 6C6, Cat. No. MABT1538, is a rat monoclonal antibody that detects human Sorting nexin-9 protein and has been tested for use in Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: A representative lot detected SNX9 in Western Blotting applications (Schobel, S., et. al. (2008). J Biol Chem. 283(21):14257-68).

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: A 1:250 dilution of this antibody detected SNX9 in HCT116 cell lysate.

Target description

~74 kDa observed; 66.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service