Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1481

Sigma-Aldrich

Anti-COL4A6 Antibody, clone 122-2

Synonym(s):

Collagen type IV alpha 6 chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

122-2, monoclonal

mol wt

calculated mol wt 163 kDa
observed mol wt ~45 kDa

purified by

using protein G

species reactivity

rat

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

Specificity

Clone 122-2 is a rat monoclonal antibody that detects Collagen type IV 6 chain (COL4A6). It targets an epitope within the C-terminal region.

Immunogen

His-tagged recombinant fragment corresponding to 231 amino acids from the C-terminal region of rat Collagen type IV 6 chain (COL4A6). It was injected into Col4a5 deficient male rats.

Application

Quality Control Testing

Evaluated by Western Blotting in GST-tagged recombinant COL4A6.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GST-tagged recombinant NC1 domain of Rat COL4A6.

Tested Applications

Immunohistochemistry Applications: A representative lot detected COL4A6 in Immunohistochemistry applications (Namba, M., et al. (2021). Sci Rep. 11(1):20836).

Western Blotting Analysis: A representative lot detected Col4a6 in Western Blotting applications (Namba, M., et al. (2021). Sci Rep. 11(1):20836).

ELISA Analysis: A representative lot detected Col4a6 in ELISA applications (Namba, M., et al. (2021). Sci Rep.11(1):20836).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Collagen type IV alpha 6 chain (UniProt: A0A0G2K601; also known as Col4a6) is encoded by the Col4a6 gene (Gene ID: 363458) in rat. The collagen super family includes over 20 different types of collagens with at least 38 distinct polypeptide chains. Of these types I, II, and III are the most abundant and are involved in the formation of fibrils. Type IV collagen forms a two-dimensional reticulum and is a major component of the basal lamina. It is a network-forming collagen that underlies epithelial and endothelial cells and functions as a barrier between tissue compartments. Type IV collagen has many binding partners and forms the backbone of the basement membrane. It also contains numerous cysteine residues that are involved in inter- and intramolecular disulfide bonding. The type IV collagen family comprises six isomeric chains, designated 1(IV) to 6(IV), each of which can form a triple helix structure with 2 other chains to generate type IV collagen network. Each type of collagen IV is a heterotrimer composed of three chains. Formation of these heterotrimers is initiated by the non-collagenous 1 (NC1) domain interactions and is accompanied by folding of the collagenous domains into triple helices. Collagen IV 6 is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to generate the mature form. Clone 122-2 specifically recognizes collagen IV 6 and reacts with the NC1 domain. (Ref.: Namba, M., et al. (2021). Sci. Rep. 11; 20836; Khoshnoodi, J., et al. (2008). Microsc. Res. Tech. 71(5); 357-370).

Physical form

Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service