Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT1349

Sigma-Aldrich

Anti-Desmoglein-2 Antibody, clone CSTEM28

Synonym(s):

Cadherin family member 5;HDGC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

CSTEM28, monoclonal

mol wt

calculated mol wt 122.29 kDa
observed mol wt ~78 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DSG2(1829)

Related Categories

General description

Desmoglein-2 (UniProt: Q14126; also known as Cadherin family member 5, HDGC) is encoded by the DSG2 (also known as CDHF5) gene (Gene ID: 1829) in human. Desmogleins are protein substances located in and on the surface of keratinocytes. They are present in specialized cell-cell adhesion structures enriched in tissues that undergo substantial mechanical stress. These proteins have been shown to be a critical factor in cell-to-cell adhesion. Desmoglein-2 is a single-pass type I membrane glycoprotein that is found in all tissues and in carcinomas. It serves as a component of intercellular desmosome junctions. It is involved in the interaction of plaque proteins and intermediate filaments mediating cell-cell adhesion. It is synthesized with a signal peptide (aa 1-23) and a propeptide (aa 24-49) that are subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 50-609), a transmembrane domain (aa 610-634), and a cytoplasmic domain (aa 635-1118). Mutations in DSG2 gene have been linked to familial type of arrhythmogenic right ventricular dysplasia that is characterized y infiltration of adipose and fibrous tissue into the right ventricle and loss of myocardial cells, resulting in ventricular and supraventricular arrhythmias. Desmoglein-2 levels are reported to be consistently upregulated in most cancers where it participates in the maintenance of intracellular junctions that prevents host immune responses. (Ref.: O Brien, CM., et al. (2017). Stem Cells. 35(3); 626-640).

Specificity

Clone CSTEM28 is a mouse monoclonal antibody that detects Desmoglein-2. It targets an epitope within the extracellular domain.

Immunogen

His-tagged recombinant fragment corresponding to the extracellular domain of human Desmoglein-2.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant human Desmoglein-2.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant Human Desmoglein-2.

Tested Applications

Flow Cytometry Analysis: A representative lot detected Desmoglein-2 in Flow Cytometry (O Brien, C.M., et al. (2017). Stem Cells. 35(3); 626-640).

Immunocytochemistry Analysis: A representative lot detected Desmoglein-2 in Immunocytochemistry (O Brien, C.M., et al. (2017). Stem Cells. 35(3); 626-640).

ELISA Analysis: A representative lot detected Desmoglein-2 in ELISA (O Brien, C.M., et al. (2017). Stem Cells. 35(3); 626-640).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Desmoglein-2 , clone CSTEM28, Cat. No. MABT1349, is a mouse monoclonal antibody that detects Desmoglein-2 and is tested for use in ELISA, Flow Cytometry, Immunocytochemistry, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service