Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABT1331

Sigma-Aldrich

Anti-ADAM 12 Antibody, clone 6E6

clone 6E6, from mouse

Synonym(s):

Disintegrin and metalloproteinase domain-containing protein 12, Meltrin-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$631.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$631.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ADAM12(8038)

General description

Disintegrin and metalloproteinase domain-containing protein 12 (UniProt: O43184, also known as ADAM 12, Meltrin-alpha) is encoded by the ADAM12 (also known as MLTN, UNQ346/PRO545) gene (Gene ID: 8038) in human. ADAM (A disintegrin and metalloprotease) family consists of multidomain cell-surface proteins that are synthesized in their proform. Four isoforms of ADAM12 have been described that are produced by alternative splicing. Isoform 1 is a single-pass transmembrane protein and isoforms 2,3, and 4 are secreted proteins. ADM12 is synthesized with a signal peptide (aa 1-28) and a propeptide (aa 29-207), which are subsequently cleaved off in the mature form. Isoform 1 shown to be expressed in placenta and in skeletal, cardiac, and smooth muscle. Isoform 2 is reported to be expressed only in placenta or in embryo and fetus. ADAM12 contains a cysteine rich domain (aa 514-649) that support cell adhesion through syndecans and triggers signaling events that lead to beta-1 integrin-dependent cell spreading. However, in carcinoma cells the binding of this domain to syndecans does not allow the integrin-mediated cell spreading. The conserved cysteine present in the cysteine-switch motif binds the catalytic zinc ion, thus inhibiting the enzyme. Dissociation of the cysteine from the zinc ion activates the enzyme. Upregulation of ADAM12 has been reported in malignant tumors. In human breast cancer cells expressing ADAM12, MMP-14 is shown to be recruited to the cell surface which results in ADAM12 activation and accelerates tumor progression. ADAM12 does not affect tumor cell proliferation, but it causes a diminution in tumor cell apoptosis and increases stromal cell apoptosis. (Ref.: Sundberg, C., et al. (2004). J. Biol. Chem. 279(49); 51601 51611; Albrechtsen, R., et al. (2013). J. Cell Sci. 126(20); 4707 4720; Kveiborg, M., et al. (2005). Cancer Res. 65(11); 4754-4761).

Specificity

Clone 6E6 detects ADAM 12-DeltaCyt. It targets an epitope with in extracellular disintegrin-cysteine-rich domains.

Immunogen

A full-length recombinant human ADAM12 secreted form expressed in HEK293 cells.

Application

Flow Cytometry Analysis: 1 µg from a representative lot detected ADAM 12 in one million ADAM 12-DeltaCyt MCF-7 cells.

Western Blotting Analysis: A representative lot detected ADAM 12 in Western Blotting applications.

Immunoprecipitation Analysis: A representative lot immunoprecipitated ADAM 12 in Immunoprecipitation applications.

Immunocytochemistry Analysis: A representative lot detected ADAM 12 in Immunocytochemistry applications.

Flow Cytometry Analysis: A representative lot detected ADAM 12 in Flow cytometry applications.
Research Category
Apoptosis & Cancer
This mouse monoclonal Anti-ADAM 12, clone 6E6, Cat. No. MABT1331, is a mouse monoclonal antibody that detects Disintegrin and metalloproteinase domain-containing protein 12 and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Western Blotting.

Quality

Evaluated by Immunocytochemistry in ADAM 12-DeltaCyt MCF-7 cells.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected ADAM 12-DeltaCyt in MCF-7 cells.

Target description

99.54 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service