Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT107

Sigma-Aldrich

Anti-Src-like-adapter Antibody/SLAP1, clone 1D9.1

clone 1D9.1, from mouse

Synonym(s):

Src-like-adapter, Src-like-adapter protein 1, SLAP-1, hSLAP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$532.00

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$532.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D9.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLA(6503)

General description

Src-like-adapter/SLAP1 protein, also known as SLAP, and encoded by the gene SLA/SLAP/SLAP1, is an adapter protein which negatively regulates T cell receptor signaling by inhibiting T cell antigen induced proliferation and works to control the positive selection and mitosis of T cells. SLAP1 also acts to connect signaling proteins such as ZAP70 with the T cell specific CBL ubiquitin ligase that plays a central role in degrading signaling proteins, and a critical role in limiting or initiating the T-cell specific- antigen-specific immunotolerance limits in T cells activation. Src-like-adapter/SLAP1 is localized to the cytoplasm and endosomes within cells. Src-like-adapter/SLAP1 is expressed in lung and fetal brain, and weakly expressed in heart, adult brain, placenta, liver, skeletal muscle, kidney and pancreas. Recent research suggest that SLAP1 may also play a role in different cancers and in modulation of receptor tyrosine kinase signaling in cancers apart from its conventional role in regulation of T cell receptor stability.

Immunogen

Linear peptide corresponding to human Src-like-adapter/SLAP1.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Src-like-adapter Antibody/SLAP1, clone 1D9.1 is validated for use in western blotting for the detection of Src-like-adapter/SLAP1.

Quality

Evaluated by Western Blotting in Human Lung tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Src-like-adapter/SLAP1 in 400 µg of human lung tissue lysate.

Target description

~38 kDa observed. The calculated molecular weight of this protein is 32 kDa; however, it may be observed at higher molecular weights due to known Post-translational modification sites.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service