Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS508

Sigma-Aldrich

Anti-GOT1 Antibody, clone 1F5.2

clone 1F5.2, from mouse

Synonym(s):

Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, cCAT, Glutamate oxaloacetate transaminase 1, Transaminase A, Aspartate aminotransferase, cytoplasmic, cAspAT, GOT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F5.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GOT1(2805)

General description

Aspartate aminotransferase, cytoplasmic (EC 2.6.2.1.; EC 2.6.2.3; UniProt P17174; also known as Aspartate aminotransferase 1, Aspartate transaminase 1, cAspAT, cCAT, Cysteine aminotransferase cytoplasmic, Cysteine transaminase cytoplasmic, Glutamate oxaloacetate transaminase 1, Growth-inhibiting protein 18, Transaminase A) is encoded by the GOT1 (also known as GIG18, ASTQTL1) gene (Gene ID 2805) in human. Aspartate aminotransferase is expressed in many types of cells and tissues and is conserved among prokaryotic and eukaryotic organisms. There exist two highly similar AST forms in mammals, the cytosolic cAST (encoded by the GOT1 gene) and the mitochondrial mAST (encoded by the GOT2 gene). Serum aspartate aminotransferase activity is largely that of cAST/GOT1, and elevated serum AST activity is associated with liver, heart, and muscle injury. AST catalyzes the reversible conversion between aspartate/alpha-ketoglutarate and oxaloacetate/glutamate, increasing oxaloacetate concentration therefore shifts the equilibrium of AST-catalzed reaction toward aspartate/alpha-ketoglutarate and away from glutatmate. Lowering blood glutamate concentration via oxaloacetate i.v. injection prompts an efflux of excess extracellular brain glutamate into the blood, which is reported to allevate the infarct size and edema in a rat model of ischemia induced by transient occlusion of the middle cerebral artery occlusion (MCAO).

Immunogen

Epitope: N-terminal half
GST-tagged Recombinant protein corresponding to the N-terminal region of human GOT1.

Application

Detect GOT1 using this mouse monoclonal Anti-GOT1 Antibody, clone 1F5.2, Cat. No. MABS508, validated for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected GOT1 in human liver hepatocytes and in human cerebellum.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected GOT1 in 10 µg of HepG2 cell lysate.

Target description

~43 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service