Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS494

Sigma-Aldrich

Anti-phospho MEKK5, (Thr845), clone PA214 Antibody

clone PA214, from rabbit

Synonym(s):

MEKK5, Apoptosis signal-regulating kinase 1, MAPK/ERK kinase kinase 5, ASK-1, MEK kinase 5, MEKK 5, ASK1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$439.00

$439.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μG
$439.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$439.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PA214, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr845)

Gene Information

human ... MAP3K5(4217)

General description

MEKK5 (MAP3K5) is a key kinase for our innate immunity and for mediating cellular signaling cascades evoked by changes in the external cellular environment. As such, MEKK5 is critical for cell fate during development, apoptosis, and cell survival as well as in modulating the host defense responses from a wide range of pathogens. MEKK5 acts as an upstream activator of the MKK/JNK signaling pathways and the p38 MAPK signaling pathways that ultimately results in activating multiple kinases such as MAP2K3/Mkk3 or MKK6/MMK7. Activation of MMEKK5 is via phosphorylation of Thr-845 within the activation loop via the recently discovered kinase MAP3K6 which forms a complex with MEKK5 and also stabilizes the protein. Expression is widespread and localized in the cytoplasmic in the perinuclear endoplasmic reticulum region.

Immunogen

linear peptide corresponding to Human phospho MEKK5 (Thr 845).

Application

Anti-phospho MEKK5, (Thr845), clone PA214 | MABS494 is an antibody against phospho MEKK5 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Evaluated by Western Blotting on HEK293A lysate
Western Blotting Analysis: 4 µg/mL dilution of this antibody detected phosphorylated MEKK5 protein in 10 µg of H2O2 treated Hek293A cell lysate

Target description

~160 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rabbit monoclonal supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service