Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS432

Sigma-Aldrich

Anti-MAP2K1/MEK1 Antibody

mouse monoclonal, 6B7.1

Synonym(s):

MAP kinase kinase 1, MEK1, MKK1, MAPKK 1, Dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAPK/ERK kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-MEK1 Antibody, clone 6B7.1, clone 6B7.1, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B7.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAP2K1(5604)

General description

Dual specificity mitogen-activated protein kinase kinase 1 (EC 2.7.12.2; UniProt Q02750; also known as ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK 1, MEK 1, MKK1) is encoded by the MAP2K1 (also known as CFC3, MAPKK1, MEK1, MKK1, PRKMK1) gene (Gene ID 5604) in human. MEK1 and the related MEK2 are dual specificity kinases of the MAP2K/STE7 kinase family. MEK1 and MEK2 share a high degree of sequence homology and perform similar functions. Upon activation by RAF, MEK1/2 catalyze the dual phosphorylation of threonine and tyrosine residues on ERK (MAPK) within a specific Thr–Glu–Tyr sequence at codons 202 to 204. This locks the kinase domain of MAPK into a catalytically competent conformation, ultimately leading to nuclear signals that regulate cellular growth and proliferation. MEK1/2 mutations have been reported to be associated with cardiofaciocutaneous syndrome 3 (CFC3) and cancers, including lung cancer, colorectal cancer, and melanomas.

Specificity

Expected to react with both the MKK1a and MKK1b spliced isoforms.

Immunogen

Epitope: C-terminal region
GST-tagged recombinant protein corresponding to the C-terminal region of human MEK1.

Application

Anti-MEK1 Antibody, clone 6B7.1 is an antibody against MEK1 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MEK1 in 10 µg of HeLa cell lysate.

Target description

~45 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service