Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS430

Sigma-Aldrich

Anti-Mcpip1, clone 2H8.2 Antibody

clone 2H8.2, from mouse

Synonym(s):

Mcpip1, Ribonuclease ZC3H12A, MCP-induced protein 1, Zinc finger CCCH domain-containing protein 12A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$476.00

$476.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$476.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$476.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H8.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZC3H12A(80149)

General description

MCP-induced protein 1 (also known as ribonuclease ZC3H12A, Zinc finger CCCH domain-containing protein 12A) is encoded by the ZC3H12A gene in human (also known as dJ423B22.1, MCPIP, MCPIP1). It belongs to the 4-member CCCH-zinc finger protein family (MCPIP1/2/3/4) that act as negative regulators in macrophage activation (PMID 18178554). MCPIP1 in particular is involved in immune response downregulation upon LPS treatment by targeting IL-6 and IL-12b with its RNase activity (PMID 22561375). MCPIP1 expresssion by MCP-1 treatment is reported to promote angiogenesis in human umbilical vein endothelial cell (HUVEC) cultures, while causing apoptotic cell death in cardiac myocytes (PMID 18364357 & 16574901).

Immunogen

GST-tagged recombinant protein corresponding to human Mcpip1.

Application

Anti-Mcpip1, clone 2H8.2 Antibody is an antibody against Mcpip1 for use in Immunohistochemistry and Immunocytochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected MCPIP1 in human kidney tissue.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MCPIP1 in 10 µg of THP-1 cell lysate.

Target description

~65 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service