Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2297

Sigma-Aldrich

Anti-TPO Antibody, clone 47

Synonym(s):

MoAb47, Thyroid peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

47, monoclonal

mol wt

calculated mol wt 102.96 kDa
observed mol wt ~120 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... TPO(7173)

Specificity

Clone 47 is a mouse monoclonal antibody that specifically detects Thyroid peroxidase.

Immunogen

Human thyroid peroxidase (hTPO) purified from thyroid microsomes by immunoaffinity chromatography.

Application

Quality Control Testing

Evaluated by Western Blotting in human thyroid tissue extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Thyroid peroxidase (TPO) in human thyroid tissue extract.

Tested Applications

ELISA Analysis: A representative lot detected Thyroid peroxidase (TPO) in ELISA applications (Ruf, J., et al. (1989). Endocrinology. 125(3):1211-8).

Radioimmunoassay: A representative lot detected Thyroid peroxidase (TPO) in Radioimmunoassay applications (Ruf, J., et al. (1989). Endocrinology. 125(3):1211-8).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Thyroid peroxidase (Uniprot: P07202; also known as TPO) is encoded by gene TPO (Gene ID: 7173) in human. It is a single-pass type I membrane protein and has an Extracellular domain (19-846 aa), a Transmembrane domain ( 847-871 aa) and a Cytoplasmic domain (872-933 aa). It is primarily found in the thyroid gland and functions in the iodination of tyrosine residues in thyroglobulin and the formation of phenoxy-ester between pairs of iodinated tyrosines to generate the thyroid hormones, thyroxine and triiodothyronine (T3 and T4). TPO is an integral part of the complex process by which the thyroid gland regulates metabolism and other vital bodily functions. Defects in this gene can cause congenital hypothyroidism, congenital goitre and thyroid hormone organification defect IIA. Moreover, TPO is a common target of autoimmune attacks in various thyroid disorders, such as Hashimoto′s thyroiditis and Graves′ disease, which can lead to disruptions in hormone synthesis and subsequent health issues.
(Ref.: Ruf, J., et al. (1989). Endocrinology. 125(3):1211-8. (PMID: 2474433).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service