Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2234

Sigma-Aldrich

Anti-ICA 512/RESP18 Antibody, clone 1348-D29-1

Synonym(s):

Receptor-type tyrosine-protein phosphatase-like N;R-PTP-N;Islet cell antigen 512;ICA 512;Islet cell autoantigen 3;PTP IA-2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$200.00
100 μG
$394.00

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$200.00
100 μG
$394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1348-D29-1, monoclonal

mol wt

calculated mol wt 105.85 kDa
observed mol wt ~110-140 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PTPRN(5798)

General description

Receptor-type tyrosine-protein phosphatase-like N (UniProt: P16849; also known as R-PTP-N, Islet cell antigen 512, ICA 512, Islet cell autoantigen 3, PTP IA-2) is encoded by the PTPRN (also known as ICA3, ICA512) gene (Gene ID: 5798) in human. ICA 512, a tyrosine phosphatase like intrinsic membrane protein, is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-34), which is subsequently cleaved off to produce the mature form that contains a lumenal domain (aa 35-575), a transmembrane domain (aa 576-600), and a cytoplasmic domain (aa 601-979). Its lumenal region contains the RESP18 homology domain that is localized in amino acids 35-131. The mature form can undergo cleavage to generate an N-terminal fragment (ICA 512-NTF), a transmembrane fragment (ICA 512-TMF), and a cleaved cytosolic fragment (ICA 512-CCF). ICA 512 expression is restricted to neuroendocrine cells and is detected in the pancreas, brain, and pituitary. It is involved in the biogenesis and turnover of insulin secretory granules (SGs) in pancreatic islet β-cells . It plays a role in insulin secretion in response to glucose. It is required for the accumulation of normal levels of insulin-containing vesicles and preventing their degradation. It plays a role in insulin secretion in response to glucose stimuli. It is also required for normal accumulation of secretory vesicles in hippocampus and pituitary. ICA 512-TMF is reported to regulate dynamics and exocytosis of insulin SGs. ICA 512-CCF is shown to translocate to the nucleus and promotes the expression of insulin and other granule-related genes. It enhances pancreatic b-cell proliferation by converging with signaling by STAT5B and STAT3. ICA 512-NTF containing the RESP18HD domain encodes for an intrinsically disordered region that acts as a condensing factor for the reversible aggregation of insulin and other β-cell proteins in a pH and zinc-regulated manner. (Ref.: Toledo, PL., et al. (2019). J. Biol. Chem. 294(21); 8564-8576; Mziaut, H., et al. (2008). Proc. Natl. Acad. Sci. USA. 105(2); 674-679; Trajkovski, M., et al. (2008). J. Biol. Chem. 283(48); 33719-33729).

Specificity

Clone 1348-D29-1 is a mouse monoclonal antibody that detects ICA 512/RESP18. It targets an epitope within the RESP18 homology domain within the N-terminal fragment.

Immunogen

GST-tagged recombinant protein fragment corresponding to the mature N-terminal fragment (without the signal peptide) of human ICA 512.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from EndoC βH1 cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected ICA 512/RESP18 in EndoC βH1 cells, but not in cells subjected to RNAi knockdown.

Tested Applications

Immunofluorescence Analysis: A representative lot detected ICA 512/RESP18 in Immunofluorescence applications (Toledo, P.L., et al. (2019). J Biol Chem. 294(21):8564-8576).

Western Blotting Analysis: A representative lot detected ICA 512/RESP18 in Western Blotting application (Toledo, P.L., et al. (2019). J Biol Chem. 294(21):8564-8576).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service