Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2219

Sigma-Aldrich

Anti-MGAT1 Antibody, clone 5F5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$249.00
100 μL
$625.00

$249.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$249.00
100 μL
$625.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$249.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5F5, monoclonal

mol wt

calculated mol wt 38.79 kDa
observed mol wt ~36 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG2κ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Mogat1(68393)

General description

2-acylglycerol O-acyltransferase 1 (UniProt: EC:2.3.1.22, Acyl-CoA:monoacylglycerol acyltransferase 1, MGAT1,Diacylglycerol acyltransferase 2-like protein 1, Monoacylglycerol O-acyltransferase 1, MGAT1) is encoded by the Moghat1 (also known as Dgat2l1) gene (Gene ID: 68393) in murine species. MGAT1 is an enzyme that catalyzes the formation of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA. Its activity is reported to be directly regulated by PPARg and it plays a role in regulating the pathway that leads to incorporation of fatty acids into triglycerides. MGAT1 activity is shown to be higher in mice fed high fat diet and by overexpression of PPARg, which leads to hepatic steatosis. Even in cultured hepatocytes, overexpression of MGAT1 is shown to increase triglyceride synthesis without affecting the expression of PPARg. The binding of PPARg to MGAT1 promoter is significantly enhanced Ob/Ob and high fat diet fed mice. MGAT1 expression is significantly reduced in PPARg knockdown murine models and knockdown of MGAT1 expression is shown to improve hepatic steatosis in high fat fed mice. It has also been shown that C3H mice that do not express PPARg and MGAT1 in the liver do not develop hepatic steatosis after being fed a high fat diet. (Ref.: Lee, YJ., et al. (2012). Proc. Natl. Acad. Sci. USA. 109(34); 13656-13661).

Specificity

Clone 5F5 is a mouse monoclonal antibody that detects Monoacylglycerol O-acyltransferase 1 (MGAT1). It targets an epitope within 13 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal half of murine 2-acylglycerol O-acyltransferase 1 (MGAT1).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells overexpressing MGAT1.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected MGAT1 in lysates from HEK293 cells transfected with mouse MGAT1, but not in untransfected cells.

Tested Applications

Western Blotting Analysis: A representative lot detected MGAT1 in Western Blotting applications (Lee, Y.J., et al. (2012). Proc. Natl. Acad. Sci. USA. 109(34):13656-61).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MGAT1, clone 5F5, Cat. No. MABS2219, is a mouse monoclonal antibody that detects 2-acylglycerol O-acyltransferase 1 (MGAT1) and is tested for use in Western Blotting,

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service